4-Methyl-isothiazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR346355-1G
APOSOR346355-1GEA 751 EUR
APOSOR346355-1G
4-Methyl-isothiazole-5-carboxylic acid
4-Methyl-isothiazole-5-carboxylic acid
Formel: C₅H₅NO₂S
Molecular Weight: 143,164 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09965921
CAS-Nummer: 15901-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR