4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-2-carboxylic acid

OR346361-1G
APOSOR346361-1GEA 136 EUR
APOSOR346361-1G
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09056814
CAS-Nummer: 307313-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR