2-Ethoxy-benzamidine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR346362-1G
APOSOR346362-1GEA 0 EUR
APOSOR346362-1G
2-Ethoxy-benzamidine hydrochloride
2-Ethoxy-benzamidine hydrochloride
Formel: C₉H₁₂N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD05663676
CAS-Nummer: 18637-00-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR