4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2,5-dicarboxylic acid diethyl ester

Lieferant: Apollo Scientific

OR346363-1G
APOSOR346363-1GEA 0 EUR
APOSOR346363-1G
4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2,5-dicarboxylic acid diethyl ester
4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2,5-dicarboxylic acid diethyl ester
Formel: C₁₃H₁₆N₂O₅
Molecular Weight: 280,277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24386147
CAS-Nummer: 459157-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR