4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 4-Oxo-4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-2-carboxylic acid

OR346364-1G
APOSOR346364-1GEA 0 EUR
APOSOR346364-1G
4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
Formel: C₈H₈N₂O₃
Molecular Weight: 180,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16658488
CAS-Nummer: 459157-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR