1-Butyl-piperidine-4-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR346369-1G
APOSOR346369-1GEA 0 EUR
APOSOR346369-1G
1-Butyl-piperidine-4-carboxylic acid ethyl ester
1-Butyl-piperidine-4-carboxylic acid ethyl ester
Formel: C₁₂H₂₃NO₂
Molecular Weight: 213,317 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16152773
CAS-Nummer: 74045-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR