(4-Chloro-3-hydroxy-phenyl)-carbamic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (4-chloro-3-hydroxyphenyl)carbamate

OR346370-500MG
APOSOR346370-500MGEA 0 EUR
APOSOR346370-500MG
(4-Chloro-3-hydroxy-phenyl)-carbamic acid tert-butyl ester
(4-Chloro-3-hydroxy-phenyl)-carbamic acid tert-butyl ester
Formel: C₁₁H₁₄ClNO₃
Molecular Weight: 243,687 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09951954
CAS-Nummer: 345893-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR