(4-Isocyanatomethyl-phenyl)-carbamic acid tert-butyl ester

Lieferant: Apollo Scientific
Achtung

OR346371-1G
APOSOR346371-1GEA 0 EUR
APOSOR346371-1G
(4-Isocyanatomethyl-phenyl)-carbamic acid tert-butyl ester
(4-Isocyanatomethyl-phenyl)-carbamic acid tert-butyl ester

When melted, may contain small amount of sediment

Formel: C₁₃H₁₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD25562989
CAS-Nummer: 202979-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR