2-(Chloromethyl)-3H-benzoimidazole-5-carbonitrile 95+%
Lieferant: Apollo Scientific
Synonyme:
2-(Chloromethyl)-1H-benzo[d]imidazole-6-carbonitrile
OR346379-1G
APOSOR346379-1GEA
751
EUR
APOSOR346379-1G
2-(Chloromethyl)-3H-benzoimidazole-5-carbonitrile 95+%
2-(Chloromethyl)-3H-benzoimidazole-5-carbonitrile
Formel:
C₉H₆ClN₃ Molecular Weight: 191,617 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234907 CAS-Nummer: 150613-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...