6-Methyl-1H-indole-2-carboxylic acid 95+%

Lieferant: Apollo Scientific

Synonyme: eIF4A3-IN-8

OR346380-1G OR346380-5G
APOSOR346380-1GEA 161 EUR
APOSOR346380-1G APOSOR346380-5G
6-Methyl-1H-indole-2-carboxylic acid 95+%
6-Methyl-1H-indole-2-carboxylic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664477
CAS-Nummer: 18474-59-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR