6-Methyl-1H-indole-2-carboxylic acid 95+%
Lieferant: Apollo Scientific
Synonyme:
eIF4A3-IN-8
OR346380-1G
OR346380-5G
APOSOR346380-1GEA
161
EUR
APOSOR346380-1G
APOSOR346380-5G
6-Methyl-1H-indole-2-carboxylic acid 95+%
6-Methyl-1H-indole-2-carboxylic acid
Formel:
C₁₀H₉NO₂ Molecular Weight: 175.184 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02664477 CAS-Nummer: 18474-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...