(Quinolin-8-ylmethyl)amine 95+%

Lieferant: Apollo Scientific

Synonyme: Quinolin-8-ylmethanamine

OR346387-1G
APOSOR346387-1GEA 318 EUR
APOSOR346387-1G
(Quinolin-8-ylmethyl)amine 95+%
(Quinolin-8-ylmethyl)amine
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08572158
CAS-Nummer: 15402-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR