5-Phenyl-[1,2,3]thiadiazole 95+%

Lieferant: Apollo Scientific

OR346390-1G
APOSOR346390-1GEA 243 EUR
APOSOR346390-1G
5-Phenyl-[1,2,3]thiadiazole 95+%
5-Phenyl-[1,2,3]thiadiazole
Formel: C₈H₆N₂S
Molecular Weight: 162,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014090
CAS-Nummer: 18212-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR