1-(4-Methoxyphenyl)-1H-pyrrole 95+%
Lieferant: Apollo Scientific
OR346408-5G
APOSOR346408-5GEA
0
EUR
APOSOR346408-5G
1-(4-Methoxyphenyl)-1H-pyrrole 95+%
1-(4-Methoxyphenyl)-1H-pyrrole
Formel:
C₁₁H₁₁NO Molecular Weight: 173.211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128350 CAS-Nummer: 5145-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...