5-(3-Chlorophenyl)-3-methylisoxazole-4-carboxaldehyde 95+%

Lieferant: Apollo Scientific

OR346423-1G
APOSOR346423-1GEA 345 EUR
APOSOR346423-1G
5-(3-Chlorophenyl)-3-methylisoxazole-4-carboxaldehyde 95+%
5-(3-Chlorophenyl)-3-methylisoxazole-4-carboxaldehyde
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15142856
CAS-Nummer: 1208081-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR