2-Bromo-5-methylnicotinic acid ethyl ester 95+%

Lieferant: Apollo Scientific

OR346424-1G
APOSOR346424-1GEA 751 EUR
APOSOR346424-1G
2-Bromo-5-methylnicotinic acid ethyl ester 95+%
2-Bromo-5-methylnicotinic acid ethyl ester
Formel: C₉H₁₀BrNO₂
Molecular Weight: 244,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16610943
CAS-Nummer: 65996-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR