4-Acetyl-1H-pyrrole-2-carboxaldehyde ≥97%

Lieferant: Apollo Scientific

OR346429-250MG
APOSOR346429-250MGEA 42.2 EUR
APOSOR346429-250MG
4-Acetyl-1H-pyrrole-2-carboxaldehyde ≥97%
4-Acetyl-1H-pyrrole-2-carboxaldehyde
Formel: C₇H₇NO₂
Molecular Weight: 137,136 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02179580
CAS-Nummer: 16168-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR