2-Chloro-4-phenyloxazole 95+%

Lieferant: Apollo Scientific
Achtung

OR346432-500MG
APOSOR346432-500MGEA 261 EUR
APOSOR346432-500MG
2-Chloro-4-phenyloxazole 95+%
2-Chloro-4-phenyloxazole
Formel: C₉H₆ClNO
Molecular Weight: 179,603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845044
CAS-Nummer: 445470-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR