2-Chloro-N,N-dimethylnicotinamide 95+%
Lieferant: Apollo Scientific
OR346435-5G
APOSOR346435-5GEA
0
EUR
APOSOR346435-5G
2-Chloro-N,N-dimethylnicotinamide 95+%
2-Chloro-N,N-dimethylnicotinamide
Formel:
C₈H₉ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD08593346 CAS-Nummer: 52943-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...