1-Benzofuran-2-carbonitrile 95+%
Lieferant: Apollo Scientific
OR346444-1G
APOSOR346444-1GEA
58
EUR
APOSOR346444-1G
1-Benzofuran-2-carbonitrile 95+%
1-Benzofuran-2-carbonitrile
Formel:
C₉H₅NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD01659779 CAS-Nummer: 41717-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...