5-Bromo-2-chloronicotinoyl chloride 95+%

Lieferant: Apollo Scientific

OR346454-1G
APOSOR346454-1GEA 393 EUR
APOSOR346454-1G
5-Bromo-2-chloronicotinoyl chloride 95+%
5-Bromo-2-chloronicotinoyl chloride
Formel: C₆H₂BrCl₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD12827961
CAS-Nummer: 78686-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR