3-Brom-4-methoxybenzoylchlorid 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Brom-4-methoxybenzoesäurechlorid

OR346455-1G
APOSOR346455-1GEA 340 EUR
APOSOR346455-1G
3-Brom-4-methoxybenzoylchlorid 95+%
3-Brom-4-methoxybenzoylchlorid
Formel: C₈H₆BrClO₂
Molecular Weight: 249,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02628379
CAS-Nummer: 81324-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR