3-Methylisothiazole-5-carboxaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

OR346460-500MG
APOSOR346460-500MGEA 359 EUR
APOSOR346460-500MG
3-Methylisothiazole-5-carboxaldehyde 95+%
3-Methylisothiazole-5-carboxaldehyde
Formel: C₅H₅NOS
Molecular Weight: 127,164 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10700240
CAS-Nummer: 88511-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR