2-Chloro-4-methanesulphonylbenzaldehyde 95+%
Lieferant: Apollo Scientific
OR346464-1G
APOSOR346464-1GEA
142
EUR
APOSOR346464-1G
2-Chloro-4-methanesulphonylbenzaldehyde 95+%
2-Chloro-4-methanesulphonylbenzaldehyde
Formel:
C₈H₇ClO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037960 CAS-Nummer: 101349-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...