5-Bromo-2-furoyl chloride 95+%

Lieferant: Apollo Scientific

Synonyme: 5-Bromofuran-2-carbonyl chloride

OR346465-1G
APOSOR346465-1GEA 380 EUR
APOSOR346465-1G
5-Bromo-2-furoyl chloride 95+%
5-Bromo-2-furoyl chloride
Formel: C₅H₂BrClO₂
Molecular Weight: 209,43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424743
CAS-Nummer: 26726-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR