(7-Chlorobenzo[b]thiophen-2-yl)methanol 95+%

Lieferant: Apollo Scientific

OR346467-1G
APOSOR346467-1GEA 369 EUR
APOSOR346467-1G
(7-Chlorobenzo[b]thiophen-2-yl)methanol 95+%
(7-Chlorobenzo[b]thiophen-2-yl)methanol
Formel: C₉H₇ClOS
Molecular Weight: 198,669 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12032281
CAS-Nummer: 1171926-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR