1,4-Dihydrobenzo[d][1,3]oxazin-2-one 95+%

Lieferant: Apollo Scientific

OR346471-1G
APOSOR346471-1GEA 345 EUR
APOSOR346471-1G
1,4-Dihydrobenzo[d][1,3]oxazin-2-one 95+%
1,4-Dihydrobenzo[d][1,3]oxazin-2-one
Formel: C₈H₇NO₂
Molecular Weight: 149,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01663880
CAS-Nummer: 13213-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR