1-Methyl-1,4-dihydrobenzo[d][1,3]oxazin-2-one 95+%

Lieferant: Apollo Scientific

OR346472-1G
APOSOR346472-1GEA 497 EUR
APOSOR346472-1G
1-Methyl-1,4-dihydrobenzo[d][1,3]oxazin-2-one 95+%
1-Methyl-1,4-dihydrobenzo[d][1,3]oxazin-2-one
Formel: C₉H₉NO₂
Molecular Weight: 163,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27937067
CAS-Nummer: 13213-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR