3,4-Dihydrobenzo[1,4]oxazin-2-one 95%
Lieferant: Apollo Scientific
OR346475-1G
APOSOR346475-1GEA
317
EUR
APOSOR346475-1G
3,4-Dihydrobenzo[1,4]oxazin-2-one 95%
3,4-Dihydrobenzo[1,4]oxazin-2-one
Formel:
C₈H₇NO₂ Molecular Weight: 149,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07369703 CAS-Nummer: 98554-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...