4-Methyl-3,4-dihydrobenzo[1,4]oxazin-2-one 95+%
Lieferant: Apollo Scientific
OR346476-1G
APOSOR346476-1GEA
420
EUR
APOSOR346476-1G
4-Methyl-3,4-dihydrobenzo[1,4]oxazin-2-one 95+%
4-Methyl-3,4-dihydrobenzo[1,4]oxazin-2-one
Formel:
C₉H₉NO₂ Molecular Weight: 163,173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27937070 CAS-Nummer: 103440-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...