3-Bromomethyl-2,5-dichloropyridine 95+%

Lieferant: Apollo Scientific

OR346477-1G
APOSOR346477-1GEA 751 EUR
APOSOR346477-1G
3-Bromomethyl-2,5-dichloropyridine 95+%
3-Bromomethyl-2,5-dichloropyridine
Formel: C₆H₄BrCl₂N
Molecular Weight: 240,913 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16607532
CAS-Nummer: 1141990-05-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR