3-(3,5-Dimethylisoxazol-4-yl)phenylamine 95+%

Lieferant: Apollo Scientific

Synonyme: 3-(3,5-Dimethylisoxazol-4-yl)aniline

OR346481-1G
APOSOR346481-1GEA 497 EUR
APOSOR346481-1G
3-(3,5-Dimethylisoxazol-4-yl)phenylamine 95+%
3-(3,5-Dimethylisoxazol-4-yl)phenylamine
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20701923
CAS-Nummer: 875628-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR