Benzofuran-7-carboxylic acid 95+%

Lieferant: Apollo Scientific

OR346485-1G
APOSOR346485-1GEA 345 EUR
APOSOR346485-1G
Benzofuran-7-carboxylic acid 95+%
Benzofuran-7-carboxylic acid
Formel: C₉H₆O₃
Molecular Weight: 162,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10000617
CAS-Nummer: 90484-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR