4-Bromo-3-methylisothiazole-5-carboxaldehyde 95+%

Lieferant: Apollo Scientific

OR346491-250MG
APOSOR346491-250MGEA 586 EUR
APOSOR346491-250MG
4-Bromo-3-methylisothiazole-5-carboxaldehyde 95+%
4-Bromo-3-methylisothiazole-5-carboxaldehyde
Formel: C₅H₄BrNOS
Molecular Weight: 206.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27937097
CAS-Nummer: 89283-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR