3-Bromo-4-methylbenzenesulphonyl chloride 95+%

Lieferant: Apollo Scientific

OR346493-1G
APOSOR346493-1GEA 145 EUR
APOSOR346493-1G
3-Bromo-4-methylbenzenesulphonyl chloride 95+%
3-Bromo-4-methylbenzenesulphonyl chloride
Formel: C₇H₆BrClO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD12922946
CAS-Nummer: 1029145-99-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR