3-Chloro-1-methoxyisoquinoline 95+%
Lieferant: Apollo Scientific
OR346496-1G
APOSOR346496-1GEA
166
EUR
APOSOR346496-1G
3-Chloro-1-methoxyisoquinoline 95+%
3-Chloro-1-methoxyisoquinoline
Formel:
C₁₀H₈ClNO Molecular Weight: 193.63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160677 CAS-Nummer: 24649-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...