3-(Bromomethyl)furan-2-carboxylic acid methyl ester 95+%

Lieferant: Apollo Scientific

OR346497-1G
APOSOR346497-1GEA 497 EUR
APOSOR346497-1G
3-(Bromomethyl)furan-2-carboxylic acid methyl ester 95+%
3-(Bromomethyl)furan-2-carboxylic acid methyl ester
Formel: C₇H₇BrO₃
Molecular Weight: 219,033 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07847868
CAS-Nummer: 23268-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR