(2-Bromo-thiophen-3-yl)methanol 95+%
Lieferant: Apollo Scientific
OR346501-1G
APOSOR346501-1GEA
217
EUR
APOSOR346501-1G
(2-Bromo-thiophen-3-yl)methanol 95+%
(2-Bromo-thiophen-3-yl)methanol
Formel:
C₅H₅BrOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025881 CAS-Nummer: 70260-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...