3-(3-Azetidinyl)pyridine dihydrochloride 95+%

Lieferant: Apollo Scientific

OR346503-250MG
APOSOR346503-250MGEA 0 EUR
APOSOR346503-250MG
3-(3-Azetidinyl)pyridine dihydrochloride 95+%
3-(3-Azetidinyl)pyridine dihydrochloride
Formel: C₈H₁₀N₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD20527356
CAS-Nummer: 1236791-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR