1,2-Dipiperidinoethane 95+%

Lieferant: Apollo Scientific

OR346505-1G
APOSOR346505-1GEA 78.3 EUR
APOSOR346505-1G
1,2-Dipiperidinoethane 95+%
1,2-Dipiperidinoethane
Formel: C₁₂H₂₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00014644
CAS-Nummer: 1932-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR