3-((E)-2-Dimethylaminovinyl)isonicotinonitrile 95+%

Lieferant: Apollo Scientific

OR346511-1G
APOSOR346511-1GEA 497 EUR
APOSOR346511-1G
3-((E)-2-Dimethylaminovinyl)isonicotinonitrile 95+%
3-((E)-2-Dimethylaminovinyl)isonicotinonitrile
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13181594
CAS-Nummer: 601514-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR