2,5-Dimethylimidazo[1,2-a]pyridine 95+%

Lieferant: Apollo Scientific

OR346512-1G
APOSOR346512-1GEA 497 EUR
APOSOR346512-1G
2,5-Dimethylimidazo[1,2-a]pyridine 95+%
2,5-Dimethylimidazo[1,2-a]pyridine
Formel: C₉H₁₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09263848
CAS-Nummer: 6188-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR