6-Methylpyridine-3-sulphonyl chloride 95+%

Lieferant: Apollo Scientific

OR346514-1G
APOSOR346514-1GEA 318 EUR
APOSOR346514-1G
6-Methylpyridine-3-sulphonyl chloride 95+%
6-Methylpyridine-3-sulphonyl chloride
Formel: C₆H₆ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11868981
CAS-Nummer: 478264-00-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR