(6-Bromopyridin-2-ylmethyl)methylamine 95+%
Lieferant: Apollo Scientific
OR346518-1G
APOSOR346518-1GEA
369
EUR
APOSOR346518-1G
(6-Bromopyridin-2-ylmethyl)methylamine 95+%
(6-Bromopyridin-2-ylmethyl)methylamine
Formel:
C₇H₉BrN₂ Molecular Weight: 201,064 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08690325 CAS-Nummer: 675109-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...