2-Methyl-5,6,7,8-tetrahydroquinazolin-6-ylamine 95+%

Lieferant: Apollo Scientific

OR346521-1G
APOSOR346521-1GEA 751 EUR
APOSOR346521-1G
2-Methyl-5,6,7,8-tetrahydroquinazolin-6-ylamine 95+%
2-Methyl-5,6,7,8-tetrahydroquinazolin-6-ylamine
Formel: C₉H₁₃N₃
Molecular Weight: 163,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08447508
CAS-Nummer: 929973-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR