O-(3-Methyl-butyl)-hydroxylamine hydrochloride 95+%

Lieferant: Apollo Scientific

OR346525-1G
APOSOR346525-1GEA 497 EUR
APOSOR346525-1G
O-(3-Methyl-butyl)-hydroxylamine hydrochloride 95+%
O-(3-Methyl-butyl)-hydroxylamine hydrochloride
Formel: C₅H₁₃NO·ClH
Molecular Weight: 139,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02249339
CAS-Nummer: 19411-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR