1-(1H-Benzoimidazol-5-yl)ethanone 95+%
Lieferant: Apollo Scientific
OR346526-1G
APOSOR346526-1GEA
0
EUR
APOSOR346526-1G
1-(1H-Benzoimidazol-5-yl)ethanone 95+%
1-(1H-Benzoimidazol-5-yl)ethanone
Formel:
C₉H₈N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09263854 CAS-Nummer: 58442-16-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...