1-(6-Phenylpyridin-3-yl)ethanone 95+%

Lieferant: Apollo Scientific

Synonyme: 1-(6-Phenyl-3-pyridinyl)ethanone

OR346534-1G
APOSOR346534-1GEA 294 EUR
APOSOR346534-1G
1-(6-Phenylpyridin-3-yl)ethanone 95+%
1-(6-Phenylpyridin-3-yl)ethanone
Formel: C₁₃H₁₁NO
Molecular Weight: 197,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12404909
CAS-Nummer: 35022-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR