3-Bromo-5-isocyanatopyridine, tech

Lieferant: Apollo Scientific

OR346535-1G
APOSOR346535-1GEA 445 EUR
APOSOR346535-1G
3-Bromo-5-isocyanatopyridine, tech
3-Bromo-5-isocyanatopyridine

May form small amount of sediment

Formel: C₆H₃BrN₂O
Molecular Weight: 199,005 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09879965
CAS-Nummer: 167951-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR