2-(Chlormethyl)chinolin 95+%

Lieferant: Apollo Scientific

OR346537-1G
APOSOR346537-1GEA 122 EUR
APOSOR346537-1G
2-(Chlormethyl)chinolin 95+%
2-(Chloromethyl)quinoline
Formel: C₁₀H₈ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD00023999
CAS-Nummer: 4377-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR